Synonym:
Fmoc-
MDL number MFCD00801269
PubChem Substance ID 24871289
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Biology, Chemical Synthesis, |
| extent of labeling | 0.4-0.6 mmol/g loading |
| matrix | polystyrene, crosslinked with 1% DVB |
| particle size | 200-400 mesh |
| storage temp. | 2-8°C |
1 g in glass bottle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?