• USA Home
  • 476714 - 3-(Ethylamino)propionitrile

EMAIL THIS PAGE TO A FRIEND
476714 Aldrich

3-(Ethylamino)propionitrile

98%

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   RUVUQOOKKGVDNN-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.433(lit.)
bp   190 °C(lit.)
density   0.883 g/mL at 25 °C(lit.)

Description

Packaging

5 mL in glass bottle

Application

3-(Ethylamino)propionitrile was used in the synthesis of 3-[ethyl- (2-naphthalen-1-yl-ethyl)-amino]-propionitrile by reacting with 1-(2-bromoethyl)naphthalene.

General description

3-(Ethylamino)propionitrile (β-ethylaminopropionitrile) is an aminonitrile. It has been reported to be synthesized by reacting ethylamine with equimolar acrylonitrile.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGIII
WGK Germany 
3
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?