• USA Home
  • 476862 - Diethylisopropylsilane

EMAIL THIS PAGE TO A FRIEND
476862 Aldrich

Diethylisopropylsilane

97%

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Other Organosilicon
InChI Key   DUPUVYJQZSLSJB-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.419(lit.)
bp   128 °C(lit.)
density   0.743 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
28.4 °F
Flash Point(C) 
-2 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?