• USA Home
  • 477036 - 4-Benzyloxybenzyl alcohol, polymer-bound

EMAIL THIS PAGE TO A FRIEND
477036 Aldrich

4-Benzyloxybenzyl alcohol, polymer-bound

200-400 mesh, extent of labeling: 1.0-1.5 mmol/g OH loading, 1 % cross-linked with divinylbenzene

Synonym: p-Alkoxybenzyl alcohol, polymer-bound, Wang resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, Wang Resins
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   1.0-1.5 mmol/g OH loading
particle size   200-400 mesh

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?