EMAIL THIS PAGE TO A FRIEND
47770 Aldrich

Fmoc-D-Phe(4-I)-OH

≥96.0% (HPLC)

Synonym: Fmoc-4-iodo-D-phenylalanine

  • CAS Number 205526-29-0

  • Empirical Formula (Hill Notation) C24H20INO4

  • Molecular Weight 513.32

  •  Beilstein Registry Number 9159551

  •  MDL number MFCD00672557

  •  PubChem Substance ID 24871406

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   LXOXXTQKKRJNNB-JOCHJYFZSA-N
assay   ≥96.0% (HPLC)
storage temp.   2-8°C

Description

Packaging

1 g in poly tube

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?