EMAIL THIS PAGE TO A FRIEND
47802 Aldrich

Fmoc-D-Phe(2-CN)-OH

purum, ≥98.0% (HPLC)

Synonym: Fmoc-2-cyano-D-phenylalanine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ACFGBBASKOIPEW-HSZRJFAPSA-N
grade   purum
assay   ≥98.0% (HPLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?