• USA Home
  • 47808 - Fmoc-D-Phe(2,4-Cl2)-OH

EMAIL THIS PAGE TO A FRIEND
47808 Aldrich

Fmoc-D-Phe(2,4-Cl2)-OH

≥98.0% (HPLC)

Synonym: Fmoc-2,4-dichloro-D-phenylalanine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   PGIMVGROPOKRNA-JOCHJYFZSA-N
assay   ≥98.0% (HPLC)
storage temp.   2-8°C

Description

Packaging

1 g in poly tube

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?