• USA Home
  • 478296 - 3-Fluoro-4-methylphenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
478296 Aldrich

3-Fluoro-4-methylphenyl isocyanate

98%

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C7-C8, Chemical Synthesis, Fluorinated Building Blocks,
InChI Key   FWRCOINKYLERQL-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.516(lit.)
bp   187-192 °C(lit.)
density   1.175 g/mL at 25 °C(lit.)

Description

Packaging

2 g in glass bottle

Application

3-Fluoro-4-methylphenyl isocyanate may be used in the synthesis of 3-fluoro analogue of 5-(acetamido-methyl)oxazolidinones.

General description

3-Fluoro-4-methylphenyl isocyanate, also known as 2-fluoro-4-isocyanato-1-methylbenzene, is an organic building block containing an isocyanate group.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?