• USA Home
  • 478350 - 2-Methyl-3-nitrophenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
478350 Aldrich

2-Methyl-3-nitrophenyl isocyanate

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   NSEABHDWHUBNCA-UHFFFAOYSA-N
assay   98%
bp   118 °C/2.8 mmHg(lit.)
mp   36-39 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3335
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?