• USA Home
  • 47974 - 3-(Fmoc-aminomethyl)benzoic acid

EMAIL THIS PAGE TO A FRIEND
47974 Aldrich

3-(Fmoc-aminomethyl)benzoic acid

≥98.0%

  • CAS Number 155369-11-2

  • Empirical Formula (Hill Notation) C23H19NO4

  • Molecular Weight 373.40

  •  Beilstein Registry Number 6825025

  •  MDL number MFCD00273463

  •  PubChem Substance ID 57651140

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   VODSFAVGEJUBHO-UHFFFAOYSA-N
assay   ≥98.0%
storage temp.   2-8°C

Description

Packaging

1 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?