EMAIL THIS PAGE TO A FRIEND
479764 Aldrich

Boc-D-1-Nal-OH

97%

Synonym: Boc-3-(1-naphthyl)-D-alanine

  • CAS Number 76932-48-4

  • Linear Formula C10H7CH2CH[NHCO2C(CH3)3]CO2H

  • Molecular Weight 315.36

  •  Beilstein Registry Number 6180114

  •  MDL number MFCD00079668

  •  PubChem Substance ID 24871646

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   KHHIGWRTNILXLL-OAHLLOKOSA-N
assay   97%
optical activity   [α]22/D +53°, c = 2.5 in methanol
mp   145-148 °C(lit.)

Description

Packaging

500 mg in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

15045 Boc-D-1-Nal-OH, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?