• USA Home
  • 479853 - N-Benzyl-4-chloro-5-sulfamoylanthranilic acid

EMAIL THIS PAGE TO A FRIEND
479853 Aldrich

N-Benzyl-4-chloro-5-sulfamoylanthranilic acid

98%

Synonym: 2-Benzylamino-4-chloro-5-sulfamoylbenzoic acid

  • CAS Number 2114-68-3

  • Linear Formula C6H5CH2NHC6H2(Cl)(SO2NH2)CO2H

  • Molecular Weight 340.78

  •  MDL number MFCD01321358

  •  PubChem Substance ID 24871655

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonamides/Sulfinamides, Sulfur Compounds More...
InChI Key   QQLNFWFHBAFHPR-UHFFFAOYSA-N
assay   98%
mp   245-248 °C(lit.)

Description

Packaging

1 g in glass bottle

General description

N-Benzyl-4-chloro-5-sulfamoylanthranilic acid, a sulfonamide derivative, is an organic building block that is employed in organic synthesis.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?