EMAIL THIS PAGE TO A FRIEND
47991 Aldrich

(+)-α-Funebrene

≥95.0% (sum of enantiomers, GC)

Synonym: (+)-1,7-Diepi-α-cedrene

  • CAS Number 50894-66-1

  • Empirical Formula (Hill Notation) C15H24

  • Molecular Weight 204.35

  •  PubChem Substance ID 24871660

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Complex Molecules
InChI Key   IRAQOCYXUMOFCW-GUIRCDHDSA-N
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D +103±1°, neat
refractive index   n20/D 1.495
bp   252 °C(lit.)
density   0.925 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Packaging

100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?