• USA Home
  • 481017 - Antimony(V) fluoride

EMAIL THIS PAGE TO A FRIEND
481017 Aldrich

Antimony(V) fluoride

99.99% trace metals basis

Synonym: Antimony pentafluoride

Purchase

Properties

Related Categories Antimony, Antimony Salts, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals,
vapor density   2.2 (vs air)
vapor pressure   10 mmHg ( 25 °C)
InChI Key   VBVBHWZYQGJZLR-UHFFFAOYSA-I
assay   99.99% trace metals basis
form   liquid
bp   148-150 °C(lit.)
mp   7 °C(lit.)
density   2.993 g/mL at 25 °C(lit.)

Description

Application

HF/SbF5 is an excellent medium for the preparation of alkylidene oxonium salts.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1732 6.1(8) / PGII
WGK Germany 
2
Protocols & Articles

Articles

Acids and Bases

Acids and bases have been used by chemists for centuries, and are among the most fundamental reagents employed in synthetic organic chemistry. This Aldrich product line ranges from Brønsted and Lewis...
Chemfiles Volume 1 Article 3

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?