• USA Home
  • 481173 - Rhodium(II) hexanoate, dimer

EMAIL THIS PAGE TO A FRIEND
481173 Aldrich

Rhodium(II) hexanoate, dimer

97%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Micro/NanoElectronics, Rhodium,
InChI Key   FNSGZOCUBHMUMB-UHFFFAOYSA-J
assay   97%
form   solid
mp   130-136 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VI9361400
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?