• USA Home
  • 481866 - 4-Methoxy-2-methylphenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
481866 Aldrich

4-Methoxy-2-methylphenyl isocyanate

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   XGRQPTXFVDRJMY-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.546(lit.)
bp   76 °C/1 mmHg(lit.)
density   1.121 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?