• USA Home
  • 482412 - Poly[dimethylsiloxane-co-methyl(3-hydroxypropyl)siloxane]-graft-poly(ethylene glycol) methyl ether

EMAIL THIS PAGE TO A FRIEND
482412 Aldrich

Poly[dimethylsiloxane-co-methyl(3-hydroxypropyl)siloxane]-graft-poly(ethylene glycol) methyl ether

viscosity ~45 cSt (lit.)

Purchase

Properties

Related Categories Bioelectronics, Copolymers, Materials Science, Micro/NanoElectronics, Microfluidics,
composition   PEG, 80 wt. %
refractive index   n20/D 1.455
viscosity   ~45 cSt(lit.)
mp   −14 °C
density   1.035 g/mL at 25 °C

Price and Availability


Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles

Articles

Kit for PDMS Surface Modification

Aldrich Materials Science, in collaboration with Clemson University and the Pennsylvania State University, has developed a Surface Modification Kit, which allows for durable hydrophilization of vario...
Keywords: Materials Science, Solvents

Micro and Nanometer Scale Photopatterning of Self-Assembled Monolayers

Graham J. Leggett* Department of Chemistry, University of Sheffield, Brook Hill, Sheffield S3 7HF, United Kingdom *Email: Graham.Leggett@shef.ac.uk
Keywords: Absorption, Adsorption, Degradations, Deposition, Microscopy, Optical microscopy, Oxidations, Polymerization reactions, Semiconductor, Spectroscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?