• USA Home
  • 483621 - Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride

EMAIL THIS PAGE TO A FRIEND
483621 Aldrich

Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride

97%

  • CAS Number 79269-71-9

  • Empirical Formula (Hill Notation) C18H26Cl2Ti

  • Molecular Weight 361.17

  •  MDL number MFCD01073774

  •  PubChem Substance ID 24871904

Purchase

Properties

Related Categories CVD and ALD Precursors by Metal, Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Micro/NanoElectronics,
InChI Key   VUWPOFFYSGYZQR-UHFFFAOYSA-L
assay   97%
mp   233 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?