• USA Home
  • 483680 - Ammonium cerium(IV) sulfate hydrate

EMAIL THIS PAGE TO A FRIEND
483680 Aldrich

Ammonium cerium(IV) sulfate hydrate

≥99.99% trace metals basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Cerium, Cerium Salts, Chemical Synthesis, Essential Chemicals,
InChI Key   KAQXQPIKRXFEPY-UHFFFAOYSA-J
assay   ≥99.99% trace metals basis
form   chunks
  granular
  powder

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?