EMAIL THIS PAGE TO A FRIEND
484350 Aldrich

Ethylferrocene

98%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Iron More...
InChI Key   XEYMPZDIHJESAH-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.601(lit.)
bp   264 °C(lit.)
density   1.256 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

• Used in the development of photosensitive GEMs with resistive electrodes manufactured via screen printing

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?