EMAIL THIS PAGE TO A FRIEND
484458 Aldrich

Niobium(V) oxide Green Alternative

mesoporous, pore size 22 Å, 99.5% trace metals basis

Synonym: Niobium pentoxide

Purchase

Properties

Related Categories Alternative Energy, Chemical Synthesis, Electrode Materials, Enabling Products, Greener Alternative Products,
InChI Key   ZKATWMILCYLAPD-UHFFFAOYSA-N
assay   99.5% trace metals basis
form   powder
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
unit cell size   3.1-3.2 nm
pore size   22 Å pore size
surface area   spec. surface area 650-800 m2/g
density   4.47 g/mL at 25 °C(lit.)

Description

Application

Promising results were obtained using niobium(V) oxide as an alternate electrode to lithium metal in advanced fuel cells.

Features and Benefits

Applied in the solid state formation of an unusual cation deficient perovskite, Ba7Nb4MoO20.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
nwg
RTECS 
QU0500000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

208515 Niobium(V) oxide, −325 mesh, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?