• USA Home
  • 493767 - (R)-(+)-4-tert-Butyl-2-oxazolidinone

EMAIL THIS PAGE TO A FRIEND
493767 Aldrich

(R)-(+)-4-tert-Butyl-2-oxazolidinone

99%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Oxazolidinone Derivatives
InChI Key   WKUHGFGTMLOSKM-YFKPBYRVSA-N
assay   99%
optical activity   [α]23/D +17°, c = 6 in ethanol
mp   119-122 °C(lit.)

Description

Application

Versatile chiral auxiliary for asymmetric synthesis.

Packaging

500 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?