• USA Home
  • 493791 - 2-Chlorobenzyl chloroformate

EMAIL THIS PAGE TO A FRIEND
493791 Aldrich

2-Chlorobenzyl chloroformate

97%

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor pressure   1.36 psi ( 20 °C)
InChI Key   GCCKNHYFOVQZRG-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.536(lit.)
bp   50 °C/100 mmHg(lit.)
density   1.339 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3277 8(6.1) / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?