Synonym: Diaspirin, Succinylsalicylic acid
CAS Number 578-19-8
Linear Formula (-CH2CO2C6H4CO2H)2
Molecular Weight 358.30
MDL number MFCD00133271
PubChem Substance ID 24872872
| Related Categories | Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters, |
| assay | 95% |
| mp | 160-165 °C(lit.) |
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?