EMAIL THIS PAGE TO A FRIEND
494291 Aldrich

Methanol

HPLC grade, ≥99.9%

Synonym: Methyl alcohol

Purchase

Properties

grade   HPLC grade
vapor density   1.11 (vs air)
vapor pressure   410 mmHg ( 50 °C)
  97.68 mmHg ( 20 °C)
InChI Key   OKKJLVBELUTLKV-UHFFFAOYSA-N
assay   ≥99.9%
autoignition temp.   725 °F
expl. lim.   36 %
availability   available only in EU
refractive index   n20/D 1.329(lit.)
bp   64.7 °C(lit.)
mp   −98 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
RTECS 
PC1400000
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?