• USA Home
  • 494704 - (R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone

EMAIL THIS PAGE TO A FRIEND
494704 Aldrich

(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Oxazolidinone Derivatives
InChI Key   QEOCTJMBYZNEJH-AWEZNQCLSA-N
assay   98%
optical activity   [α]20/D +50°, c = 1 in chloroform
mp   145-148 °C(lit.)

Description

Legal Information

Licensed under patent no. 5939554

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?