• USA Home
  • 496057 - (S)-(−)-1,2,3,4-Tetrahydro-α,α-diphenyl-3-isoquinolinemethanol

EMAIL THIS PAGE TO A FRIEND
496057 Aldrich

(S)-(−)-1,2,3,4-Tetrahydro-α,α-diphenyl-3-isoquinolinemethanol

97%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Isoquinolines More...
InChI Key   RHDBEHVNWCDLDP-NRFANRHFSA-N
assay   97%
optical activity   [α]22/D −100°, c = 1 in methanol
mp   104-107 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?