• USA Home
  • 496227 - 4-Bromo-2-chlorophenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
496227 Aldrich

4-Bromo-2-chlorophenyl isocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   QWWBEOZNMPWYQR-UHFFFAOYSA-N
assay   97%
mp   71-76 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN3077 - class 9 - PG 3 - DOT/IATA UN3335 - Environmentally hazardous substances, solid, n.o.s., HI: all (not BR)
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?