• USA Home
  • 496510 - Bis[4-(vinyloxy)butyl] (4-methyl-1,3-phenylene)biscarbamate

EMAIL THIS PAGE TO A FRIEND
496510 Aldrich

Bis[4-(vinyloxy)butyl] (4-methyl-1,3-phenylene)biscarbamate

97%

Synonym: VEctomer® 4220

Purchase

Properties

Related Categories Materials Science, Monomers, Polymer Science, Vinyl Ethers
InChI Key   RGENPSRZOHMDOK-UHFFFAOYSA-N
assay   97%
composition   Bis[4-(vinyloxy)butyl] (2-methyl-1,3-phenylene)biscarbamate isomer, 20%
mp   53 °C(lit.)

Description

Legal Information

Product of Morflex, Inc.

VEctomer is a registered trademark of Vertellus Specialties, Inc.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
298 °F
Flash Point(C) 
148 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?