EMAIL THIS PAGE TO A FRIEND
496545 Aldrich

Graphite

rod, L 150 mm, diam. 3 mm, 99.995% trace metals basis

Purchase

Properties

Related Categories Carbon, Electronic Chemicals, Graphite and Carbon-Based Materials, Materials Science, Metal and Ceramic Science,
assay   99.995% trace metals basis
form   rod
L   150 mm
diam.   3 mm
mp   3652-3697 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MD9659600

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?