• USA Home
  • 497703 - [(S,S)-Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]titanium(IV) (S)-1,1′-binaphthyl-2,2′-diolate

EMAIL THIS PAGE TO A FRIEND
497703 Aldrich

[(S,S)-Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]titanium(IV) (S)-1,1′-binaphthyl-2,2′-diolate

  • CAS Number 83417-93-0

  • Empirical Formula (Hill Notation) C40H38O2Ti

  • Molecular Weight 598.60

  •  MDL number MFCD01863624

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Other Chiral Catalysts
InChI Key   ZVPRKHQZEULVLC-UHFFFAOYSA-L
optical activity   [α]20/D +700°, c = 0.07 in methylene chloride
mp   245 °C (dec.)(lit.)

Description

Application

Reactant for:
• Dehydrocoupling reactions

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?