• USA Home
  • 49810 - DL-Glyceraldehyde Dimer

EMAIL THIS PAGE TO A FRIEND
49810 Aldrich

DL-Glyceraldehyde Dimer

≥97.0% (T)

Synonym: 3,6-Dihydroxy-1,4-dioxane-2,5-dimethanol

  • CAS Number 23147-59-3

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein Registry Number 635844

  •  MDL number MFCD00070528

  •  PubChem Substance ID 57651225

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C1 to C6, Carbohydrate Chemistry, Carbohydrate Derivatives,
InChI Key   NGNVWCSFFIVLAR-UHFFFAOYSA-N
assay   ≥97.0% (T)
impurities   ≤1% Free acid
  ~1% water
mp   136-140 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?