EMAIL THIS PAGE TO A FRIEND
50340 Aldrich

Gly-Leu-Tyr

≥98.0% (TLC)

Synonym: Glycyl-L-leucyl-L-tyrosine

  • CAS Number 4306-24-5

  • Empirical Formula (Hill Notation) C17H25N3O5

  • Molecular Weight 351.40

  •  Beilstein Registry Number 2179546

  •  EC Number 224-319-1

  •  MDL number MFCD00043036

  •  PubChem Substance ID 57651214

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Poly Amino Acids More...
InChI Key   AFWYPMDMDYCKMD-KBPBESRZSA-N
assay   ≥98.0% (TLC)
optical activity   [α]20/D −40±3°, c = 1% in 1 M HCl

Description

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 50340.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?