• USA Home
  • 511196 - Phenyl trifluoromethyl sulfide

EMAIL THIS PAGE TO A FRIEND
511196 Aldrich

Phenyl trifluoromethyl sulfide

98%

Synonym: Trifluoromethyl phenyl thioether, [(Trifluoromethyl)thio]benzene

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Fluorination Reagents, Organic Building Blocks, Sulfides/Disulfides,
InChI Key   YQQKTCBMKQQOSM-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.466(lit.)
bp   142 °C(lit.)
density   1.249 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

Phenyl trifluoromethyl sulfide may be used in the preparation of:
• sulfoxides, dithiane dioxide, and dithiolane dioxide via iron-catalyzed oxidation of sulfides, dithiane, or dithiolane with hydrogen peroxide
• benzophenone hydrazone derivatives
• trifluoro- and difluoromethylsilanes by reductive coupling of fluoromethyl sulfones, sulfoxides and sulfides with chlorosilanes

General description

Phenyl trifluoromethyl sulfide (PTFMS) is an aryl trifluoromethyl sulfide. PTFMS with (triethylgermyl)sodium forms a reagent system that has been employed for the trifluoromethylation of certain imines. The reaction between OH radical and PTFMS has been analyzed using pulse radiolysis technique and quantum chemical calculations.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
98.6 °F
Flash Point(C) 
37 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?