• USA Home
  • 512583 - Bis(butylcyclopentadienyl)tungsten(IV) dibromide

EMAIL THIS PAGE TO A FRIEND
512583 Aldrich

Bis(butylcyclopentadienyl)tungsten(IV) dibromide

98%

  • CAS Number 90023-17-9

  • Empirical Formula (Hill Notation) C18H26Br2W

  • Molecular Weight 586.05

  •  MDL number MFCD10567402

  •  PubChem Substance ID 24873644

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Tungsten More...
InChI Key   CDSRJYVHRGDLTI-UHFFFAOYSA-L
assay   98%
mp   83-87 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?