• USA Home
  • 512591 - Bis(butylcyclopentadienyl)tungsten(IV) diiodide

EMAIL THIS PAGE TO A FRIEND
512591 Aldrich

Bis(butylcyclopentadienyl)tungsten(IV) diiodide

97%

Purchase

Properties

Related Categories CVD and ALD Precursors by Metal, Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Micro/NanoElectronics,
InChI Key   FJVPRUNMUOEFSU-UHFFFAOYSA-L
assay   97%
mp   112-116 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?