• USA Home
  • 514705 - Ethyl 4′-methoxy-4-biphenylcarboxylate

EMAIL THIS PAGE TO A FRIEND
514705 Aldrich

Ethyl 4′-methoxy-4-biphenylcarboxylate

90%

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   LGFDQZCUDVWAIQ-UHFFFAOYSA-N
assay   90%
mp   102-108 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?