• USA Home
  • 514861 - Bis[4-(vinyloxy)butyl]terephthalate

EMAIL THIS PAGE TO A FRIEND
514861 Aldrich

Bis[4-(vinyloxy)butyl]terephthalate

97%

Synonym: VEctomer® 4050

Purchase

Properties

Related Categories Materials Science, Monomers, Polymer Science, Vinyl Ethers
InChI Key   HMNFSPVCKZFHGZ-UHFFFAOYSA-N
assay   97%
mp   44-46 °C(lit.)

Description

Legal Information

Product of Morflex, Inc.

VEctomer is a registered trademark of Vertellus Specialties, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
280.4 °F
Flash Point(C) 
138 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?