• USA Home
  • 515663 - Benzyl alcohol, polymer-bound

EMAIL THIS PAGE TO A FRIEND
515663 Aldrich

Benzyl alcohol, polymer-bound

70-90 mesh, extent of labeling: 0.5-1.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym: (Hydroxymethyl)polystyrene, Benzyl alcohol polymer-bound, Hydroxymethyl polystyrene, Polystyrene cross-linked with divinylbenzene, hydroxymethylated

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   0.5-1.0 mmol/g loading
particle size   70-90 mesh

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?