EMAIL THIS PAGE TO A FRIEND
51591 Aldrich

Boc-Gly-PAM resin

extent of labeling: ~0.7 mmol/g loading

Synonym: Boc-glycine bound to PAM resin

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Polymer Supports for SPPS, Resins for Boc-/Bzl-SPPS More...
extent of labeling   ~0.7 mmol/g loading
particle size   100-200 mesh
storage temp.   2-8°C

Description

Packaging

5 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?