• USA Home
  • 516236 - Sulfonyl chloride, polymer-bound

EMAIL THIS PAGE TO A FRIEND
516236 Aldrich

Sulfonyl chloride, polymer-bound

100-200 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Purchase

Properties

Related Categories Chemical Synthesis, Electrophilic, Scavengers, Solid Supported Synthesis, Specialty Synthesis More...
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   1.0-2.0 mmol/g loading
particle size   100-200 mesh

Description

Application

Reacts with nucleophiles

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

498211 Sulfonyl chloride, polymer-bound, 100-200 mesh, extent of labeling: 1.5-2.0 mmol/g loading, 1 % cross-linked

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?