• USA Home
  • 51747 - (4R)-Boc-4-benzyl-Pyr-OBzl

EMAIL THIS PAGE TO A FRIEND
51747 Aldrich

(4R)-Boc-4-benzyl-Pyr-OBzl

≥96.0% (HPLC)

Synonym: (4R)-Boc-4-benzyl-L-pyroglutamic acid benzyl ester, Benzyl (2S,4R)-1-Boc-4-benzyl-5-oxo-2-pyrrolidinecarboxylate

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Pyroglutamine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   LMILJTUPJYRGKN-UXHICEINSA-N
assay   ≥96.0% (HPLC)

Description

Other Notes

Synthesis of serine protease inhibitors

Packaging

500 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Cyclic Aryl-Substitued Amino Acid Derivatives

Aromatic a-substituted proline analogs and g-substituted pyroglutamates are valuable building blocks in both medicinal and peptide chemistry. Through Suzuki coupling technology, they can be used as s...
ChemFiles Volume 1 Article 1
Keywords: Building blocks, Peptidomimetics, Suzuki coupling

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?