• USA Home
  • 518514 - Dodecyl chloroformate

EMAIL THIS PAGE TO A FRIEND
518514 Aldrich

Dodecyl chloroformate

98%

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor pressure   1.33 mmHg ( 20 °C)
InChI Key   AFPOMDNRTZLRMD-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.442(lit.)
density   0.922 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?