• USA Home
  • 518662 - N-Hydroxymethyl-3,4,5,6-tetrahydrophthalimide

EMAIL THIS PAGE TO A FRIEND
518662 Aldrich

N-Hydroxymethyl-3,4,5,6-tetrahydrophthalimide

98%

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Cyclic Imides, Organic Building Blocks More...
InChI Key   QQHOVRKETYPQHY-UHFFFAOYSA-N
assay   98%
mp   81-85 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?