• USA Home
  • 518832 - N,N-Diethyldiethylenetriamine

EMAIL THIS PAGE TO A FRIEND
518832 Aldrich

N,N-Diethyldiethylenetriamine

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   CEFDTSBDWYXVHY-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.459(lit.)
bp   225-230 °C(lit.)
density   0.865 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN2 8 / PGII
WGK Germany 
3
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?