EMAIL THIS PAGE TO A FRIEND
51995 Aldrich

N-Heptylmethylamine

≥96.0% (GC)

Synonym: N-Methylheptylamine

  • CAS Number 36343-05-2

  • Linear Formula CH3(CH2)5CH2NHCH3

  • Molecular Weight 129.24

  •  Beilstein Registry Number 906719

  •  EC Number 252-987-4

  •  MDL number MFCD00039980

  •  PubChem Substance ID 57651339

Purchase

Properties

Related Categories Amines, Building Blocks, C8, Chemical Synthesis, Nitrogen Compounds,
InChI Key   LTGYRKOQQQWWAF-UHFFFAOYSA-N
assay   ≥96.0% (GC)
refractive index   n20/D 1.423
bp   53-54 °C/6 mmHg(lit.)
density   0.768 g/mL at 20 °C(lit.)

Description

Packaging

10 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
125.6 °F
Flash Point(C) 
52 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?