• USA Home
  • 521035 - PSS-(3-Cyclohexen-1-yl)-Heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
521035 Aldrich

PSS-(3-Cyclohexen-1-yl)-Heptacyclopentyl substituted

Synonym: 1-(3-Cyclohexen-1-yl)-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Cyclohexenyl-POSS®

  • CAS Number 307496-29-3

  • Empirical Formula (Hill Notation) C41H72O12Si8

  • Molecular Weight 981.69

  •  MDL number MFCD02093735

Purchase

Properties

Related Categories Completely Condensed, Materials Science, Nanomaterials, Silsesquioxanes: POSS Nanohybrids
InChI Key   VHADFQZVTXSMFQ-UHFFFAOYSA-N
mp   >350 °C(lit.)

Description

Packaging

5 g in glass bottle

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?