• USA Home
  • 52136 - Boc-β-Lys(Z)-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
52136 Aldrich

Boc-β-Lys(Z)-OH (dicyclohexylammonium) salt

≥98.0% (TLC)

Synonym: Nβ-Boc-Nω-Z-L-β-lysine (dicyclohexylammonium) salt, Boc-L-β-Homoorn(Z)-OH (dicyclohexylammonium) salt

  • CAS Number 401792-89-0

  • Empirical Formula (Hill Notation) C19H28N2O6 · C12H23N

  • Molecular Weight 561.75

  •  MDL number MFCD11656033

  •  PubChem Substance ID 24874202

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   DIIXPGUGJXWBQW-RSAXXLAASA-N
assay   ≥98.0% (TLC)

Description

Packaging

1 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?