• USA Home
  • 522295 - PSS-[2-[(4-Chloromethyl)phenyl]ethyl]-Heptacyclopentyl substituted

EMAIL THIS PAGE TO A FRIEND
522295 Aldrich

PSS-[2-[(4-Chloromethyl)phenyl]ethyl]-Heptacyclopentyl substituted

97%

Synonym: (Chloromethyl)phenylethyl-POSS®, 1-[2-[(Chloromethyl)phenyl]ethyl]-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, mixture of isomers

  • Empirical Formula (Hill Notation) C44H73ClO12Si8

  • Molecular Weight 1054.18

  •  MDL number MFCD02093749

Purchase

Properties

Related Categories Completely Condensed, Materials Science, Nanomaterials, Silsesquioxanes: POSS Nanohybrids
assay   97%
mp   230 °C (dec.)(lit.)

Description

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

673323 PSS-[2-[(Chloromethyl)phenyl]ethyl]-Heptaisobutyl substituted, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?