• USA Home
  • 522384 - 2,4-Diamino-6-mercaptopyrimidine sulfate salt

EMAIL THIS PAGE TO A FRIEND
522384 Aldrich

2,4-Diamino-6-mercaptopyrimidine sulfate salt

Synonym: 2,6-Diamino-4-pyrimidinethiol sulfate

  • CAS Number 307496-37-3

  • Empirical Formula (Hill Notation) C4H6N4S · H2SO4

  • Molecular Weight 240.26

  •  MDL number MFCD01863745

  •  PubChem Substance ID 24874270

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
InChI Key   FVSVUQJNAZQSFX-UHFFFAOYSA-N
impurities   <2 mol sulfate
mp   >300 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?